Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloropyridine, 99%

Click to view available options
Quantity:
10g
50g
626-60-8
C5H4ClN
113.544
MFCD00006375
PWRBCZZQRRPXAB-UHFFFAOYSA-N
pyridine, 3-chloro,m-chloropyridine,unii-1m13huc1p4,ccris 1715,chloropyridine,b-chloropyridine,3-chloro pyridine,3-chloro-pyridine,3-chloropyridine,3-chloranylpyridine
12287
3-chloropyridine
C1=CC(=CN=C1)Cl
626-60-8
C5H4ClN
113.544
MFCD00006375
PWRBCZZQRRPXAB-UHFFFAOYSA-N
pyridine, 3-chloro,m-chloropyridine,unii-1m13huc1p4,ccris 1715,chloropyridine,b-chloropyridine,3-chloro pyridine,3-chloro-pyridine,3-chloropyridine,3-chloranylpyridine
12287
3-chloropyridine
C1=CC(=CN=C1)Cl
Applications
3-Chloropyridine is used as a pharmaceutical intermediate. It can be used to produce 3-chloro-pyridine-1-oxide.

Solubility
Soluble in water 10 g/L.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Boiling Point 147°C to 149°C
Flash Point 53°C (127°F)
Quantity 10g
UN Number UN1993
Beilstein 105744
Formula Weight 113.55
Percent Purity 99%
Chemical Name or Material 3-Chloropyridine

GHS H Statement
H314-H318-H226-H302-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : US6125000

TSCA : TSCA