Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Chloropyridine 95.0+%, TCI America™
Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Color | Yellow |
Boiling Point | 148°C |
Quantity | 100 g |
Formula Weight | 113.54 |
Percent Purity | ≥95.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 3-Chloropyridine |
Safety and Handling
RTECSNumber : US6125000
TSCA : Yes