Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chlorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Click to view available options
Quantity:
5g
25g
100g
2039-85-2
C8H7Cl
138.594
MFCD00000598
BOVQCIDBZXNFEJ-UHFFFAOYSA-N
3-chlorostyrene,1-chloro-3-vinylbenzene,m-chlorostyrene,benzene, 1-chloro-3-ethenyl,styrene, m-chloro,unii-11k8g759hc,1-chloro-3-vinyl-benzene,styrene, 3-chloro,meta-chlorostyrene,zlchem 318
14905
1-chloro-3-ethenylbenzene
C=CC1=CC(=CC=C1)Cl
2039-85-2
C8H7Cl
138.594
MFCD00000598
BOVQCIDBZXNFEJ-UHFFFAOYSA-N
3-chlorostyrene,1-chloro-3-vinylbenzene,m-chlorostyrene,benzene, 1-chloro-3-ethenyl,styrene, m-chloro,unii-11k8g759hc,1-chloro-3-vinyl-benzene,styrene, 3-chloro,meta-chlorostyrene,zlchem 318
14905
1-chloro-3-ethenylbenzene
C=CC1=CC(=CC=C1)Cl
Applications
It has been identified as a potential candidate for an electron resist to be used in direct device fabrication.

Solubility
Not miscible or difficult to mix in water. Soluble in ether, and ethanol

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
Boiling Point 188°C to 189°C
Quantity 25g
Flash Point 67°C (153°F)
Odor Characteristic
Beilstein 2038489
Formula Weight 138.6
Percent Purity 98%
Chemical Name or Material 3-Chlorostyrene, stabilized with 0.1% 4-tert-butylcatechol

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Recommended Storage : 2° to 8°C