Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Chlorosulfonyl)benzoic acid, 97%

Click to view available options
Quantity:
5g
25g
100g
4025-64-3
C7H5ClO4S
220.623
MFCD00024877
LMRKXSDOAFUINK-UHFFFAOYSA-N
3-chlorosulfonyl benzoic acid,benzoic acid, 3-chlorosulfonyl,3-carboxybenzenesulfonyl chloride,m-chlorosulfonylbenzoic acid,3-carboxyphenylsulfonyl chloride,benzoic acid, m-chlorosulfonyl,3-chlorosulphonyl benzoic acid,3-carboxylbenzenesulfonyl chloride,3-chlorosulfonyl-benzoic acid,m-chlorosulfonyl benzoic acid
77641
3-chlorosulfonylbenzoic acid
C1=CC(=CC(=C1)S(=O)(=O)Cl)C(=O)O
4025-64-3
C7H5ClO4S
220.623
MFCD00024877
LMRKXSDOAFUINK-UHFFFAOYSA-N
3-chlorosulfonyl benzoic acid,benzoic acid, 3-chlorosulfonyl,3-carboxybenzenesulfonyl chloride,m-chlorosulfonylbenzoic acid,3-carboxyphenylsulfonyl chloride,benzoic acid, m-chlorosulfonyl,3-chlorosulphonyl benzoic acid,3-carboxylbenzenesulfonyl chloride,3-chlorosulfonyl-benzoic acid,m-chlorosulfonyl benzoic acid
77641
3-chlorosulfonylbenzoic acid
C1=CC(=CC(=C1)S(=O)(=O)Cl)C(=O)O
Quantity 5g
Melting Point 133°C to 138°C
UN Number UN3261
Beilstein 2645465
Formula Weight 220.63
Percent Purity 97%
Chemical Name or Material 3-(Chlorosulfonyl)benzoic acid

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA