Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chlorothiophene, 98%, may contain up to 2% DMF

Click to view available options
Quantity:
5g
25g
100g
17249-80-8
C4H3ClS
118.578
MFCD00043887
QUBJDMPBDURTJT-UHFFFAOYSA-N
thiophene, 3-chloro,3-chloro thiophene,3-thienyl chloride,3-chlorthiophen,3-chloro-thiophene,3-chloranylthiophene,pubchem5477,acmc-1cdmv,3-chlorothiophene,ksc175g2h
87017
3-chlorothiophene
C1=CSC=C1Cl
17249-80-8
C4H3ClS
118.578
MFCD00043887
QUBJDMPBDURTJT-UHFFFAOYSA-N
thiophene, 3-chloro,3-chloro thiophene,3-thienyl chloride,3-chlorthiophen,3-chloro-thiophene,3-chloranylthiophene,pubchem5477,acmc-1cdmv,3-chlorothiophene,ksc175g2h
87017
3-chlorothiophene
C1=CSC=C1Cl
Refractive Index 1.552
Density 1.276
Quantity 5g
Boiling Point 137°C to 139°C
Flash Point 37°C (98°F)
Odor Unpleasant
UN Number UN1992
Beilstein 105255
Formula Weight 118.59
Percent Purity 98%
Chemical Name or Material 3-Chlorothiophene
Assay may contain up to 2% DMF
Show More Show Less

GHS H Statement
H331-H360-H318-H302-H312-H315-H335
Toxic if inhaled.
May damage fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H318-H335-H350-H360D-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen