Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chlorothiophene, 98%, may contain up to 2% DMF

Click to view available options
Quantity:
5g
25g
100g
17249-80-8
C4H3ClS
118.578
MFCD00043887
QUBJDMPBDURTJT-UHFFFAOYSA-N
thiophene, 3-chloro,3-chloro thiophene,3-thienyl chloride,3-chlorthiophen,3-chloro-thiophene,3-chloranylthiophene,pubchem5477,acmc-1cdmv,3-chlorothiophene,ksc175g2h
87017
3-chlorothiophene
C1=CSC=C1Cl
17249-80-8
C4H3ClS
118.578
MFCD00043887
QUBJDMPBDURTJT-UHFFFAOYSA-N
thiophene, 3-chloro,3-chloro thiophene,3-thienyl chloride,3-chlorthiophen,3-chloro-thiophene,3-chloranylthiophene,pubchem5477,acmc-1cdmv,3-chlorothiophene,ksc175g2h
87017
3-chlorothiophene
C1=CSC=C1Cl
Boiling Point 137°C to 139°C
Odor Unpleasant
Flash Point 37°C (98°F)
Quantity 100g
UN Number UN1992
Beilstein 105255
Formula Weight 118.59
Percent Purity 98%
Chemical Name or Material 3-Chlorothiophene
Assay may contain up to 2% DMF

GHS H Statement
H331-H360-H318-H302-H312-H315-H335
Toxic if inhaled.
May damage fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.