Alfa Aesar™ 3-Coumaranone, 97%

Manufacturer: Alfa Aesar™ A1020203

Quantity 1g

 View more versions of this product

Catalog No. AAA1020203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7169-34-8
    Molecular Formula C8H6O2
    Formula Weight 134.13
    MDL Number MFCD00051810
    Beilstein 115296
    Synonym 2H-Benzofuran-3-one; 2,3-Dihydrobenzofuran-3-one
    Chemical Name or Material 3-Coumaranone

    Specifications

    Formula Weight 134.13
    Synonym 2H-Benzofuran-3-one; 2,3-Dihydrobenzofuran-3-one
    Quantity 1g
    Molecular Formula C8H6O2
    MDL Number MFCD00051810
    CAS 7169-34-8
    Beilstein 115296
    Melting Point 97° to 102°C
    Recommended Storage −30°C to −10°C
    Chemical Name or Material 3-Coumaranone
    Assay 97%

    Description

    Applications
    3-Coumaranone is used in the aurones, [2-benzylidenebenzofuran-3(2H)-ones] by reacting with appropriate aldehyde. It is involved in the condensation reaction with aldehydes in the presence of morpholine acetate to prepare substituted 2-(arylidene)benzofuran-3(ZH)-ones. In Horner-Wadsworth-Emmons reaction, it reacts with diethyl¬ cyanomethyl¬phosphonate to give 3-(cyanomethyl)benzofurans, which is an intermediate in the synthesis of antihyperglycemic agents.

    Solubility
    Slightly miscible with water.

    Notes
    Light sensitive. Incompatible with strong oxidizing agents.

    −30°C to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit