Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-CPMT, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
14008-79-8
C21H25Cl2NO
378.337
FXKCQWIUITUJFU-AMUQYKJESA-N
3α-[(4-Chlorophenyl)phenylmethoxy] tropane hydrochloride
124081001
(1R,5S)-3-[(R)-(4-chlorophenyl)-phenylmethoxy]-8-methyl-8-azabicyclo[3.2.1]octane;hydrochloride
CN1C2CCC1CC(C2)OC(C3=CC=CC=C3)C4=CC=C(C=C4)Cl.Cl
14008-79-8
C21H25Cl2NO
378.337
FXKCQWIUITUJFU-AMUQYKJESA-N
3α-[(4-Chlorophenyl)phenylmethoxy] tropane hydrochloride
124081001
(1R,5S)-3-[(R)-(4-chlorophenyl)-phenylmethoxy]-8-methyl-8-azabicyclo[3.2.1]octane;hydrochloride
CN1C2CCC1CC(C2)OC(C3=CC=CC=C3)C4=CC=C(C=C4)Cl.Cl

Dopamine uptake inhibitor

This cocaine analog binds with high affinity to the dopamine transporter (Ki = 30 nM) and blocks re-uptake of dopamine. However, it is a weak psychomotor stimulant and does not substitute for cocaine in addicted rats.
Quantity 50mg
Formula Weight 378.34
Percent Purity >99%
Chemical Name or Material 3-CPMT

Recommended Storage : Store at Room Temperature