Alfa Aesar™ 3-Cyano-1-propylboronic acid pinacol ester, 96%

Manufacturer: Alfa Aesar™ H2712614

Quantity 25g

 View more versions of this product

Catalog No. AAH2712614

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 238088-16-9
    Molecular Formula C10H18BNO2
    Formula Weight 195.07
    MDL Number MFCD09953480
    UN Number UN3276
    Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)butyronitrile
    Chemical Name or Material 3-Cyano-1-propylboronic acid pinacol ester

    Specifications

    Formula Weight 195.07
    Molecular Formula C10H18BNO2
    MDL Number MFCD09953480
    UN Number UN3276
    Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)butyronitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Quantity 25g
    CAS 238088-16-9
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3-Cyano-1-propylboronic acid pinacol ester
    Assay 96%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit