Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Cyano-2-fluorobenzeneboronic acid pinacol ester, 96%

Click to view available options
Quantity:
250mg
1g
1218791-15-1
C13H15BFNO2
247.076
MFCD11506358
BDIHSSFYJYXWRV-UHFFFAOYSA-N
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,3-cyano-2-fluorophenylboronic acid pinacol ester,3-cyano-2-fluorophenylboronic acid,pinacol ester,3-cyano-2-fluorophenylboronic acid, pinacol ester,3-cyano-2-fluoro benzene boronic acid, pinacol ester,2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,benzonitrile, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
53217116
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C#N)F
1218791-15-1
C13H15BFNO2
247.076
MFCD11506358
BDIHSSFYJYXWRV-UHFFFAOYSA-N
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,3-cyano-2-fluorophenylboronic acid pinacol ester,3-cyano-2-fluorophenylboronic acid,pinacol ester,3-cyano-2-fluorophenylboronic acid, pinacol ester,3-cyano-2-fluoro benzene boronic acid, pinacol ester,2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,benzonitrile, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
53217116
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C#N)F
Quantity 250mg
Formula Weight 247.07
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3-Cyano-2-fluorobenzeneboronic acid pinacol ester

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures