Alfa Aesar™ 3-Cyano-2-methoxypyridine, 97+%

Manufacturer: Alfa Aesar™ H52373MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52373MD

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7254-34-4
    Molecular Formula C7H6N2O
    Formula Weight 134.14
    MDL Number MFCD07363804
    Synonym 2-Methoxypyridine-3-carbonitrile; 2-Methoxynicotinonitrile
    Chemical Name or Material 3-Cyano-2-methoxypyridine

    Specifications

    Formula Weight 134.14
    Molecular Formula C7H6N2O
    MDL Number MFCD07363804
    CAS 7254-34-4
    Synonym 2-Methoxypyridine-3-carbonitrile; 2-Methoxynicotinonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 250mg
    Chemical Name or Material 3-Cyano-2-methoxypyridine
    Assay ≥97%

    Description

    Applications
    3-Cyano-2-methoxypyridine is used as an organic chemical synthesis intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit