Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-4-fluorophenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
775351-57-0
C13H15BFNO2
247.08
RYJOVQGRIOURGT-UHFFFAOYSA-N
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,3-cyano-4-fluorophenylboronic acid, pinacol ester,3-cyano-4-fluorophenylboronic acid pinacol ester,benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-cyano-4-fluorobenzeneboronic acid pinacol ester,3-cyano-4-fluorophenyl-boronic acid pinacol ester,2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile,2-fluoro-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzonitrile,2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile
11402361
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C#N
775351-57-0
C13H15BFNO2
247.08
RYJOVQGRIOURGT-UHFFFAOYSA-N
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,3-cyano-4-fluorophenylboronic acid, pinacol ester,3-cyano-4-fluorophenylboronic acid pinacol ester,benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-cyano-4-fluorobenzeneboronic acid pinacol ester,3-cyano-4-fluorophenyl-boronic acid pinacol ester,2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile,2-fluoro-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzonitrile,2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile
11402361
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C#N
Quantity 5g
Melting Point 75.0°C to 76.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 247.08
Percent Purity 97%
Chemical Name or Material 3-Cyano-4-fluorophenylboronic acid pinacol ester

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.

GHS P Statement
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a posi

GHS Signal Word: Danger