Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Cyano-6-methyl-2-pyridone, 98%

Manufacturer:  Alfa Aesar™ B2018830

Quantity 250g

 View more versions of this product

Catalog No. AAB2018830


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

Applications
3-Cyano-6-methyl-2(1H)-pyridinone is a reactant used in the synthesis of Milrinone (M344680), a selective phosphodiesterase inhibitor with vasodilating and positive inotropic activity. 3-It has also been used in the preparation of the N3 ft.-pyridyl thiamine, a potent in vitro thiamine antagonist.

Solubility
Soluble in water at 20°C, 2.38 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

4241-27-4
134.14
120412
3-Cyano-6-methyl-2-pyridone
C7H6N2O
MFCD00006013
3-Cyano-2-hydroxy-6-methylpyridine; 2-Hydroxy-6-methylnicotinonitrile

Specifications

≈298°C (dec.)
4241-27-4
134.14
120412
3-Cyano-6-methyl-2-pyridone
250g
C7H6N2O
MFCD00006013
3-Cyano-2-hydroxy-6-methylpyridine; 2-Hydroxy-6-methylnicotinonitrile
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit