Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-7-ethoxycoumarin, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
117620-77-6
C12H9NO3
215.208
YAFGHMIAFYQSCF-UHFFFAOYSA-N
3-cyano-7-ethoxycoumarin,7-ethoxy-2-oxo-2h-chromene-3-carbonitrile,7-ethoxycoumarin-3-carbonitrile,2h-1-benzopyran-3-carbonitrile, 7-ethoxy-2-oxo,2h-1-benzopyran-3-carbonitrile,7-ethoxy-2-oxo,3-cetc,acmc-1by0u,7-ethoxy-2-oxo-chromene-3-carbonitrile,3-cyano-7-ethoxycoumarin hplc,7-ethoxy-2-oxo-1-benzopyran-3-carbonitrile
164045
7-ethoxy-2-oxochromene-3-carbonitrile
CCOC1=CC2=C(C=C1)C=C(C(=O)O2)C#N
This item is not returnable. View return policy
117620-77-6
C12H9NO3
215.208
YAFGHMIAFYQSCF-UHFFFAOYSA-N
3-cyano-7-ethoxycoumarin,7-ethoxy-2-oxo-2h-chromene-3-carbonitrile,7-ethoxycoumarin-3-carbonitrile,2h-1-benzopyran-3-carbonitrile, 7-ethoxy-2-oxo,2h-1-benzopyran-3-carbonitrile,7-ethoxy-2-oxo,3-cetc,acmc-1by0u,7-ethoxy-2-oxo-chromene-3-carbonitrile,3-cyano-7-ethoxycoumarin hplc,7-ethoxy-2-oxo-1-benzopyran-3-carbonitrile
164045
7-ethoxy-2-oxochromene-3-carbonitrile
CCOC1=CC2=C(C=C1)C=C(C(=O)O2)C#N

Fluorescent cytochrome P450 substrate

Fluorescent P450 substrate (excitation/emission wavelengths = 408/455 nm); metabolized to cyano-hydroxycoumarin.
Quantity 10mg
Formula Weight 215.2
Percent Purity >98%
Chemical Name or Material 3-Cyano-7-ethoxycoumarin

Recommended Storage : Store at +4°C