Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-7-ethoxycoumarin, Tocris Bioscience™

Catalog Number 481550
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
117620-77-6
C12H9NO3
215.208
YAFGHMIAFYQSCF-UHFFFAOYSA-N
3-cyano-7-ethoxycoumarin, 7-ethoxy-2-oxo-2h-chromene-3-carbonitrile, 7-ethoxycoumarin-3-carbonitrile, 2h-1-benzopyran-3-carbonitrile, 7-ethoxy-2-oxo, 2h-1-benzopyran-3-carbonitrile,7-ethoxy-2-oxo, 3-cetc, acmc-1by0u, 7-ethoxy-2-oxo-chromene-3-carbonitrile, 3-cyano-7-ethoxycoumarin hplc, 7-ethoxy-2-oxo-1-benzopyran-3-carbonitrile
164045
7-ethoxy-2-oxochromene-3-carbonitrile
CCOC1=CC2=C(C=C1)C=C(C(=O)O2)C#N
This item is not returnable. View return policy
117620-77-6
C12H9NO3
215.208
YAFGHMIAFYQSCF-UHFFFAOYSA-N
3-cyano-7-ethoxycoumarin, 7-ethoxy-2-oxo-2h-chromene-3-carbonitrile, 7-ethoxycoumarin-3-carbonitrile, 2h-1-benzopyran-3-carbonitrile, 7-ethoxy-2-oxo, 2h-1-benzopyran-3-carbonitrile,7-ethoxy-2-oxo, 3-cetc, acmc-1by0u, 7-ethoxy-2-oxo-chromene-3-carbonitrile, 3-cyano-7-ethoxycoumarin hplc, 7-ethoxy-2-oxo-1-benzopyran-3-carbonitrile
164045
7-ethoxy-2-oxochromene-3-carbonitrile
CCOC1=CC2=C(C=C1)C=C(C(=O)O2)C#N

Fluorescent cytochrome P450 substrate

Fluorescent P450 substrate (excitation/emission wavelengths = 408/455 nm); metabolized to cyano-hydroxycoumarin.
Quantity 50 mg
Formula Weight 215.2
Percent Purity >98%
Chemical Name or Material 3-Cyano-7-ethoxycoumarin
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit