Learn More
Aat BioquestSupplier Diversity Partner 3-Cyano-7-hydroxycoumarin *CAS 19088-73-4* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 40
3-Cyano-7-hydroxycoumarin is used as a calibration standard for 3-cyano-7-hydroxycoumarin-based enzyme substrates. It has pH-dependent fluorescence.
Catalog No. 50-195-2678
Due to product restrictions, please to purchase or view availability for this product.