Alfa Aesar™ 3-Cyano-L-phenylalanine, 98%

Manufacturer: Alfa Aesar™ H27474MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27474MD

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 57213-48-6
    Molecular Formula C10H10N2O2
    Formula Weight 190.2
    MDL Number MFCD01860885
    UN Number UN3439
    Synonym H-Phe(3-CN)-OH
    Chemical Name or Material 3-Cyano-L-phenylalanine

    Specifications

    Formula Weight 190.2
    Molecular Formula C10H10N2O2
    MDL Number MFCD01860885
    CAS 57213-48-6
    UN Number UN3439
    Synonym H-Phe(3-CN)-OH
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Quantity 250mg
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 3-Cyano-L-phenylalanine
    Assay 98%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit