Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Cyanobenzeneboronic acid pinacol ester, 97%

Click to view available options
Quantity:
1g
5g
214360-46-0
C13H16BNO2
229.086
MFCD03789262
FIGQEPXOSAFKTA-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,3-cyanophenylboronic acid pinacol ester,3-cyanophenylboronic acid, pinacol ester,benzonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-cyanobenzeneboronic acid pinacol ester,3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,pubchem15603,3-cyanophenylboronicacidpinacolester,3-cyanobenzeneboronic acid, pinacol ester,3-cyanophenylboronic acid pinacol ester
2760590
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C#N
214360-46-0
C13H16BNO2
229.086
MFCD03789262
FIGQEPXOSAFKTA-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,3-cyanophenylboronic acid pinacol ester,3-cyanophenylboronic acid, pinacol ester,benzonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-cyanobenzeneboronic acid pinacol ester,3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,pubchem15603,3-cyanophenylboronicacidpinacolester,3-cyanobenzeneboronic acid, pinacol ester,3-cyanophenylboronic acid pinacol ester
2760590
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C#N
Quantity 1g
Melting Point 78°C to 82°C (decomposition)
Color White
Formula Weight 229.09
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Cyanobenzeneboronic acid pinacol ester

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning