Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanobenzoyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1711-11-1
C8H4ClNO
165.58
MFCD00011545
RPESZQVUWMFBEO-UHFFFAOYSA-N
3-cyanobenzoylchloride,m-cyanobenzoyl chloride,unii-m7zy44hlss,m7zy44hlss,benzoyl chloride, 3-cyano,3-cyanobenzoic acid chloride,3-cyanobenzoyl-chloride,pubchem10466,acmc-20an5i,3-cyano-benzoyl chloride
308483
3-cyanobenzoyl chloride
C1=CC(=CC(=C1)C(=O)Cl)C#N
1711-11-1
C8H4ClNO
165.58
MFCD00011545
RPESZQVUWMFBEO-UHFFFAOYSA-N
3-cyanobenzoylchloride,m-cyanobenzoyl chloride,unii-m7zy44hlss,m7zy44hlss,benzoyl chloride, 3-cyano,3-cyanobenzoic acid chloride,3-cyanobenzoyl-chloride,pubchem10466,acmc-20an5i,3-cyano-benzoyl chloride
308483
3-cyanobenzoyl chloride
C1=CC(=CC(=C1)C(=O)Cl)C#N
Boiling Point 125°C to 129°C (11.0mmHg)
Flash Point 113°C
Color Gray or White to Yellow
Melting Point 38°C to 44°C
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula NCC6H4COCl
Formula Weight 165.58
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Percent Purity 98%
Chemical Name or Material 3-Cyanobenzoyl chloride
Show More Show Less

GHS H Statement
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger