Alfa Aesar™ 3-(Cyanomethyl)benzeneboronic acid pinacol ester, 96%

Manufacturer: Alfa Aesar™ H5261803

Quantity 1g

 View more versions of this product

Catalog No. AAH5261803

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 396131-82-1
    Molecular Formula C14H18BNO2
    Formula Weight 243.11
    MDL Number MFCD02179458
    Chemical Name or Material 3-(Cyanomethyl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 243.11
    Molecular Formula C14H18BNO2
    MDL Number MFCD02179458
    CAS 396131-82-1
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 44° to 48°C
    Quantity 1g
    Chemical Name or Material 3-(Cyanomethyl)benzeneboronic acid pinacol ester
    Assay 96%

    Description

    Applications
    3-(Cyanomethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit