Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanophenylacetic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1878-71-3
C9H7NO2
161.16
ZNXMNKGBHYVNNE-UHFFFAOYSA-N
2-3-cyanophenyl acetic acid,3-cyanophenylacetic acid,3-cyanophenyl acetic acid,m-cyanophenylacetic acid,benzeneacetic acid, 3-cyano,3-cyano-phenyl-acetic acid,acmc-20a9gi,3-cyano phenyl acetic acid,benzeneacetic acid,3-cyano
1519397
2-(3-cyanophenyl)acetic acid
C1=CC(=CC(=C1)CC(=O)O)C#N
1878-71-3
C9H7NO2
161.16
ZNXMNKGBHYVNNE-UHFFFAOYSA-N
2-3-cyanophenyl acetic acid,3-cyanophenylacetic acid,3-cyanophenyl acetic acid,m-cyanophenylacetic acid,benzeneacetic acid, 3-cyano,3-cyano-phenyl-acetic acid,acmc-20a9gi,3-cyano phenyl acetic acid,benzeneacetic acid,3-cyano
1519397
2-(3-cyanophenyl)acetic acid
C1=CC(=CC(=C1)CC(=O)O)C#N
Melting Point 117°C to 118°C
Color White to Brown
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 161.16
Physical Form Powder or Crystalline Powder
Percent Purity 95%
Chemical Name or Material 3-Cyanophenylacetic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning