Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Cyclohexyl-L-alanine, 95%

Click to view available options
Quantity:
1g
5g
27527-05-5
C9H17NO2
171.24
MFCD00153484
ORQXBVXKBGUSBA-QMMMGPOBSA-N
s-2-amino-3-cyclohexylpropanoic acid,l-cyclohexylalanine,h-cha-oh,2s-2-amino-3-cyclohexylpropanoic acid,3-cyclohexyl-l-alanine,l-3-cyclohexylalanine,beta-cyclohexyl-alanine,s-2-amino-3-cyclohexylpropionic acid,l-beta-cyclohexylalanine,3-cyclohexylalanine
712421
(2S)-2-amino-3-cyclohexylpropanoic acid
C1CCC(CC1)CC(C(=O)O)N
27527-05-5
C9H17NO2
171.24
MFCD00153484
ORQXBVXKBGUSBA-QMMMGPOBSA-N
s-2-amino-3-cyclohexylpropanoic acid,l-cyclohexylalanine,h-cha-oh,2s-2-amino-3-cyclohexylpropanoic acid,3-cyclohexyl-l-alanine,l-3-cyclohexylalanine,beta-cyclohexyl-alanine,s-2-amino-3-cyclohexylpropionic acid,l-beta-cyclohexylalanine,3-cyclohexylalanine
712421
(2S)-2-amino-3-cyclohexylpropanoic acid
C1CCC(CC1)CC(C(=O)O)N
Formula Weight 171.24
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material 3-Cyclohexyl-L-alanine

storageNote1 : 2° to 8°C