Alfa Aesar™ 3-Cyclohexylpropionic acid, 98+%

Manufacturer: Alfa Aesar™ B236130B

Quantity 1000g

 View more versions of this product

Catalog No. AAB236130B

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 701-97-3
    Molecular Formula C9H16O2
    Formula Weight 156.23
    MDL Number MFCD00001527
    Beilstein 2042799
    Synonym 3-Cyclohexanepropionic acid
    Chemical Name or Material 3-Cyclohexylpropionic acid

    Specifications

    Formula Weight 156.23
    TSCA TSCA
    Molecular Formula C9H16O2
    MDL Number MFCD00001527
    Beilstein 2042799
    Synonym 3-Cyclohexanepropionic acid
    Health Hazard 2 GHS H Statement
    H301-H315-H319
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Flash Point 130°C (266°F)
    Odor Unpleasant
    Boiling Point 276°C
    Melting Point 14° to 17°C
    Quantity 1000g
    CAS 701-97-3
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3-Cyclohexylpropionic acid
    Assay ≥98%

    Description

    Applications
    3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.

    Solubility
    Soluble in water 10g/ L.

    Notes
    Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases.

    GHS H Statement
    H301-H315-H319
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit