Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclopentene-1-carboxylic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
7686-77-3
C6H8O2
112.128
MFCD00101764
XVSYDLITVYBCBD-UHFFFAOYSA-N
3-cyclopentene-1-carboxylic acid,cyclopent-3-enecarboxylic acid,3-cyclopentenecarboxylic acid,3-cyclopentene-1-carhoxylic acid,cyclopent-3-ene carboxylic acid,cyclopentene-3-carboxylic acid,ghl.pd_mitscher_leg0.243,3-cyclopentene-1-carboxylicacid,acmc-1bh22,ksc377a8b
853756
cyclopent-3-ene-1-carboxylic acid
C1C=CCC1C(=O)O
7686-77-3
C6H8O2
112.128
MFCD00101764
XVSYDLITVYBCBD-UHFFFAOYSA-N
3-cyclopentene-1-carboxylic acid,cyclopent-3-enecarboxylic acid,3-cyclopentenecarboxylic acid,3-cyclopentene-1-carhoxylic acid,cyclopent-3-ene carboxylic acid,cyclopentene-3-carboxylic acid,ghl.pd_mitscher_leg0.243,3-cyclopentene-1-carboxylicacid,acmc-1bh22,ksc377a8b
853756
cyclopent-3-ene-1-carboxylic acid
C1C=CCC1C(=O)O
Color Colorless to Yellow
Packaging Glass bottle
Density 1.0840g/mL
Quantity 1g
Refractive Index 1.4680 to 1.4720 (20°C, 589nm)
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 112.13
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-Cyclopentene-1-carboxylic acid
Show More Show Less