Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclopentyl-1-propanol, 99%, Acros Organics

Click to view available options
Quantity:
1g
767-05-5
C8H16O
128.215
MFCD00001393
IBMXMCXCSPGCDQ-UHFFFAOYSA-N
3-cyclopentyl-1-propanol,cyclopentanepropanol,3-cyclopentylpropanol,cyclopentyl-1-propanol,cyclopentane-1-propanol,3-cyclopentyl propanol,acmc-1bg9n,3-cyclopentyl-propan-1-ol,1-cyclopentyl-3-hydroxypropane
69842
3-cyclopentylpropan-1-ol
C1CCC(C1)CCCO
767-05-5
C8H16O
128.215
MFCD00001393
IBMXMCXCSPGCDQ-UHFFFAOYSA-N
3-cyclopentyl-1-propanol,cyclopentanepropanol,3-cyclopentylpropanol,cyclopentyl-1-propanol,cyclopentane-1-propanol,3-cyclopentyl propanol,acmc-1bg9n,3-cyclopentyl-propan-1-ol,1-cyclopentyl-3-hydroxypropane
69842
3-cyclopentylpropan-1-ol
C1CCC(C1)CCCO
Quantity 1g
Refractive Index 1.4580 to 1.4600 (20°C, 589nm)
Infrared Spectrum Authentic
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C5H9(CH2)3OH
Formula Weight 128.21
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 3-Cyclopentyl-1-propanol, 99%
Show More Show Less