Learn More
R&D Systems™ 3-D Culture Cell Harvest Buffers
R&D Systems™ 3-D Culture Cell Harvest Buffers provide an optimized and standardized solution for the isolation and normalization of cell lysates from 3-D Culture Matrix™ BME or Laminin I for subsequent biochemical analysis.
Manufacturer:  R&D Systems™ 3448020CH
Catalog No. 34-480-20CH
Add to cart
This item is not returnable. View return policy
Description
Description
  • Do not use past expiration date
  • 3D Cultures exhibit cellular behaviors and morphologies similar to those seen in vivo, however, the adaptation of these models for studying biochemical processes has been impeded by the challenge of separating intact cells from extra-cellular proteins comprising the hydrogel.
  • Contains reagents that may be harmful if swallowed, or come in contact with skin or eyes. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice
  • Proprietary formulation containing sodium citrate, EDTA, and glycerol
Specifications
Specifications
100 mL