Alfa Aesar™ 3-Decanol, 97%

Manufacturer: Alfa Aesar™ B2414106

Quantity 5g

 View more versions of this product

Catalog No. AAB2414106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1565-81-7
    Molecular Formula C10H22O
    Formula Weight 158.29
    MDL Number MFCD00021956
    Beilstein 1719658
    Synonym Ethyl n-heptyl carbinol
    Chemical Name or Material 3-Decanol

    Specifications

    Formula Weight 158.29
    TSCA TSCA
    Flash Point 99°C (210°F)
    Molecular Formula C10H22O
    Boiling Point 213°C
    MDL Number MFCD00021956
    CAS 1565-81-7
    Beilstein 1719658
    Quantity 5g
    Synonym Ethyl n-heptyl carbinol
    Chemical Name or Material 3-Decanol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit