Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Decanol, 97%

Click to view available options
Quantity:
5g
25g
100g
1565-81-7
C10H22O
158.285
MFCD00021956
ICEQLCZWZXUUIJ-UHFFFAOYSA-N
3-decanol,heptyl ethyl carbinol,ethyl heptyl carbinol,3-decanol, r,3-decanol, s,fema no. 3605,3-decyl alcohol,ethylheptylcarbinol,deca-8-yl alcohol,acmc-20mxdt
519158
decan-3-ol
CCCCCCCC(CC)O
1565-81-7
C10H22O
158.285
MFCD00021956
ICEQLCZWZXUUIJ-UHFFFAOYSA-N
3-decanol,heptyl ethyl carbinol,ethyl heptyl carbinol,3-decanol, r,3-decanol, s,fema no. 3605,3-decyl alcohol,ethylheptylcarbinol,deca-8-yl alcohol,acmc-20mxdt
519158
decan-3-ol
CCCCCCCC(CC)O
Flash Point 99°C (210°F)
Boiling Point 213°C
Quantity 100g
Beilstein 1719658
Formula Weight 158.29
Percent Purity 97%
Chemical Name or Material 3-Decanol