Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dimethylamino-1-propanol 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
3179-63-3
C5H13NO
103.16
MFCD00002945
PYSGFFTXMUWEOT-UHFFFAOYSA-N
3-dimethylamino-1-propanol,3-dimethylamino propan-1-ol,1-propanol, 3-dimethylamino,dimethylaminopropanol,n,n-dimethylpropanolamine,3-dimethylamino propanol,dimethylpropanolamine,3-dimethlyamino propanol,3-n,n-dimethylamino propanol
76646
3-(dimethylamino)propan-1-ol
CN(C)CCCO
3179-63-3
C5H13NO
103.16
MFCD00002945
PYSGFFTXMUWEOT-UHFFFAOYSA-N
3-dimethylamino-1-propanol,3-dimethylamino propan-1-ol,1-propanol, 3-dimethylamino,dimethylaminopropanol,n,n-dimethylpropanolamine,3-dimethylamino propanol,dimethylpropanolamine,3-dimethlyamino propanol,3-n,n-dimethylamino propanol
76646
3-(dimethylamino)propan-1-ol
CN(C)CCCO
Viscosity 6.85 mPa.s (20°C)
Specific Gravity 0.886
Boiling Point 159.0°C
Infrared Spectrum Authentic
Refractive Index 1.4350 to 1.4370
Density 0.8860g/mL
Packaging Glass bottle
Flash Point 54°C
Quantity 500g
Color Colorless to Yellow
Melting Point -35.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2N(CH2)3OH
Beilstein 04, I, 433
Solubility Information Solubility in water: soluble
Formula Weight 103.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Dimethylamino-1-propanol, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-659-2

RTECSNumber : UB3155000

TSCA : TSCA