Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)benzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number p-4228614
Click to view available options
Quantity:
1 g
5 g
23501-93-1
C9H13NO
151.21
MFCD00017342
YTUXZVSDDHCTBZ-UHFFFAOYSA-N
3-dimethylamino phenyl methanol, 3-dimethylamino benzyl alcohol, 3-dimethylaminobenzyl alcohol, m-dimethylamino benzyl alcohol, benzenemethanol,3-dimethylamino, 3-dimethylamino phenyl methan-1-ol, benzyl alcohol, m-dimethylamino, benzenemethanol, 3-dimethylamino
90133
[3-(dimethylamino)phenyl]methanol
CN(C)C1=CC=CC(=C1)CO
23501-93-1
C9H13NO
151.21
MFCD00017342
YTUXZVSDDHCTBZ-UHFFFAOYSA-N
3-dimethylamino phenyl methanol, 3-dimethylamino benzyl alcohol, 3-dimethylaminobenzyl alcohol, m-dimethylamino benzyl alcohol, benzenemethanol,3-dimethylamino, 3-dimethylamino phenyl methan-1-ol, benzyl alcohol, m-dimethylamino, benzenemethanol, 3-dimethylamino
90133
[3-(dimethylamino)phenyl]methanol
CN(C)C1=CC=CC(=C1)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 109.0°C to 110.0°C (0.1 Torr)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 151.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)benzyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 245-700-9

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit