Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dimethylaminobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number AC115750250
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
99-64-9
C9H11NO2
165.19
MFCD00002497
NEGFNJRAUMCZMY-UHFFFAOYSA-N
3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid
66837
3-(dimethylamino)benzoic acid
CN(C)C1=CC=CC(=C1)C(=O)O
99-64-9
C9H11NO2
165.19
MFCD00002497
NEGFNJRAUMCZMY-UHFFFAOYSA-N
3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid
66837
3-(dimethylamino)benzoic acid
CN(C)C1=CC=CC(=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 150.0°C to 153.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula (CH3)2NC6H4CO2H
Quantity 25 g
Beilstein 14, 392
Solubility Information Solubility in water: insoluble.
Formula Weight 165.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Dimethylaminobenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 202-775-2
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit