Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Dimethylaminopropyl chloride, 97%

Click to view available options
Quantity:
1g
5g
25g
109-54-6
C5H12ClN
121.608
MFCD00044496
NYYRRBOMNHUCLB-UHFFFAOYSA-N
3-chloro-1-n,n-dimethyl propylamine,dimethylaminopropyl chloride,3-chloropropyl dimethylamine,3-chloropropyl dimethyl amine,1-dimethylamino-3-chloropropane,1-propanamine, 3-chloro-n,n-dimethyl,n-3-chloropropyl dimethylamine,3-chloro-propyl-dimethyl-amine,3-dimethylaminopropyl chloride,3-dimethylaminopropylchloride
66960
3-chloro-N,N-dimethylpropan-1-amine
CN(C)CCCCl
109-54-6
C5H12ClN
121.608
MFCD00044496
NYYRRBOMNHUCLB-UHFFFAOYSA-N
3-chloro-1-n,n-dimethyl propylamine,dimethylaminopropyl chloride,3-chloropropyl dimethylamine,3-chloropropyl dimethyl amine,1-dimethylamino-3-chloropropane,1-propanamine, 3-chloro-n,n-dimethyl,n-3-chloropropyl dimethylamine,3-chloro-propyl-dimethyl-amine,3-dimethylaminopropyl chloride,3-dimethylaminopropylchloride
66960
3-chloro-N,N-dimethylpropan-1-amine
CN(C)CCCCl
Applications
It is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Sensitivity Air Sensitive
Refractive Index 1.4320
Melting Point 168°C
Quantity 25g
Density 0.929g/mL
Color Colorless
Linear Formula Cl(CH2)3N(CH3)2
Beilstein 605294
Formula Weight 121.61
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Dimethylaminopropyl chloride
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C