Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dimethylaminopropylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
5L
Packaging:
Glass bottle
Plastic drum
109-55-7
C5H14N2
102.18
MFCD00008216
IUNMPGNGSSIWFP-UHFFFAOYSA-N
3-dimethylaminopropylamine,3-dimethylamino propylamine,3-dimethylamino-1-propylamine,n,n-dimethyl-1,3-propanediamine,1,3-propanediamine, n,n-dimethyl,n,n-dimethyltrimethylenediamine,3-aminopropyldimethylamine,n,n-dimethylpropane-1,3-diamine,3-aminopropyl dimethylamine,dimethylaminopropylamine
7993
N',N'-dimethylpropane-1,3-diamine
CN(C)CCCN
109-55-7
C5H14N2
102.18
MFCD00008216
IUNMPGNGSSIWFP-UHFFFAOYSA-N
3-dimethylaminopropylamine,3-dimethylamino propylamine,3-dimethylamino-1-propylamine,n,n-dimethyl-1,3-propanediamine,1,3-propanediamine, n,n-dimethyl,n,n-dimethyltrimethylenediamine,3-aminopropyldimethylamine,n,n-dimethylpropane-1,3-diamine,3-aminopropyl dimethylamine,dimethylaminopropylamine
7993
N',N'-dimethylpropane-1,3-diamine
CN(C)CCCN
Specific Gravity 0.81
Viscosity 1.6 mPa.s (20°C)
Infrared Spectrum Authentic
Boiling Point 133.0°C
Packaging Glass bottle
Refractive Index 1.4340 to 1.4360
Quantity 1L
Water 0.5% max.
Color Undesignated
Flash Point 32°C
Melting Point -60.0°C
Density 0.8100g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2N(CH2)3NH2
Beilstein 04, III, 554
Fieser 01,274
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 102.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Dimethylaminopropylamine
Show More Show Less

GHS P Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger