Learn More
Alfa Aesar™ 3-Ethoxy-2,6-difluorobenzylamine, 97%
Manufacturer:  Alfa Aesar™ H31587MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH31587MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
1092461-29-4
187.19
UN2735
C9H11F2NO
MFCD11519371
3-Ethoxy-2,6-difluorobenzylamine
Specifications
250mg
C9H11F2NO
MFCD11519371
3-Ethoxy-2,6-difluorobenzylamine
1092461-29-4
187.19
UN2735
97%
Safety and Handling
SDS & Safety Documents
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.