Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxy-2-cyclohexen-1-one, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
5323-87-5
C8H12O2
140.18
MFCD00001580
JWCFJPLIRVYENQ-UHFFFAOYSA-N
3-ethoxy-2-cyclohexen-1-one,3-ethoxycyclohex-2-enone,2-cyclohexen-1-one, 3-ethoxy,3-ethoxy-2-cyclohexenone,3-ethoxy-2-cyclohexene-1-one,3-ethoxycyclohex-2-ene-1-one,1-ethoxycyclohexene-3-one,acmc-209l4y,3-ethoxy-2-cyclohexen-one,3-ethoxy-cyclohex-2-enone
79216
3-ethoxycyclohex-2-en-1-one
CCOC1=CC(=O)CCC1
5323-87-5
C8H12O2
140.18
MFCD00001580
JWCFJPLIRVYENQ-UHFFFAOYSA-N
3-ethoxy-2-cyclohexen-1-one,3-ethoxycyclohex-2-enone,2-cyclohexen-1-one, 3-ethoxy,3-ethoxy-2-cyclohexenone,3-ethoxy-2-cyclohexene-1-one,3-ethoxycyclohex-2-ene-1-one,1-ethoxycyclohexene-3-one,acmc-209l4y,3-ethoxy-2-cyclohexen-one,3-ethoxy-cyclohex-2-enone
79216
3-ethoxycyclohex-2-en-1-one
CCOC1=CC(=O)CCC1
Packaging Glass bottle
Refractive Index 1.5030 to 1.505
Boiling Point 76°C to 78°C (1.0mmHg)
Flash Point 107°C
Specific Gravity 1.04
Color White to Peach
Quantity 100g
Density 1.0400g/mL
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C2H5OC6H7(=O)
Solubility Information Solubility in water: slightly soluble
Formula Weight 140.18
Physical Form Crystalline Powder
Percent Purity ≥98.5% (GC)
Chemical Name or Material 3-Ethoxy-2-cyclohexen-1-one
Show More Show Less