Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Ethoxy-4-methoxybenzaldehyde, 99%

Manufacturer:  Alfa Aesar™ B2476222

Quantity 100g

 View more versions of this product

Catalog No. AAB2476222


  Check Availability
Add to cart

Specifications

Description

Applications
3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Air sensitive. Store in a tightly sealed container. Store away from oxidizing agents, air.
Description & Specifications

Chemical Identifiers

1131-52-8
180.2
3-Ethoxy-p-anisaldehyde
C10H12O3
MFCD00010128
3-Ethoxy-4-methoxybenzaldehyde

Specifications

51° to 53°C
1131-52-8
180.2
3-Ethoxy-p-anisaldehyde
99%
100g
C10H12O3
MFCD00010128
3-Ethoxy-4-methoxybenzaldehyde
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit