Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
Reactant involved in suzuki-miyaura cross-coupling reactions, aerobic oxidative cross-coupling, rhodium-catalyzed addition reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant involved in suzuki-miyaura cross-coupling reactions, aerobic oxidative cross-coupling, rhodium-catalyzed addition reactions.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Reactant involved in suzuki-miyaura cross-coupling reactions, aerobic oxidative cross-coupling, rhodium-catalyzed addition reactions.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Specifications
Melting Point | 173°C to 176°C |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 233.98 |
Percent Purity | 98% |
Chemical Name or Material | 3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction