Alfa Aesar™ 3'-Ethoxyacetanilide, 99+%

Manufacturer: Alfa Aesar™ B2533906

Quantity 5g

 View more versions of this product

Catalog No. AAB2533906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 591-33-3
    Molecular Formula C10H13NO2
    Formula Weight 179.22
    MDL Number MFCD00061110
    Synonym 3-Acetophenetidide
    Chemical Name or Material 3'-Ethoxyacetanilide

    Specifications

    Formula Weight 179.22
    MDL Number MFCD00061110
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 95° to 98°C
    TSCA TSCA
    Molecular Formula C10H13NO2
    CAS 591-33-3
    Synonym 3-Acetophenetidide
    Quantity 5g
    RTECS Number AM4300000
    Chemical Name or Material 3'-Ethoxyacetanilide
    Assay ≥99%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit