Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Ethoxybenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
90555-66-1
C8H11BO3
165.983
MFCD00274219
CHCWUTJYLUBETR-UHFFFAOYSA-N
3-ethoxyphenyl boronic acid,3-ethoxybenzeneboronic acid,3-ethoxyphenylboronicacid,3-ethoxyphenyl boranediol,m-ethoxyphenylboronic acid,boronic acid, 3-ethoxyphenyl,pubchem1835,acmc-209r6q,3-ethoxy-phenylboronic acid
2773920
(3-ethoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OCC)(O)O
90555-66-1
C8H11BO3
165.983
MFCD00274219
CHCWUTJYLUBETR-UHFFFAOYSA-N
3-ethoxyphenyl boronic acid,3-ethoxybenzeneboronic acid,3-ethoxyphenylboronicacid,3-ethoxyphenyl boranediol,m-ethoxyphenylboronic acid,boronic acid, 3-ethoxyphenyl,pubchem1835,acmc-209r6q,3-ethoxy-phenylboronic acid
2773920
(3-ethoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OCC)(O)O
Quantity 5g
Melting Point 142°C to 146°C
Formula Weight 165.99
Percent Purity 98%
Chemical Name or Material 3-Ethoxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning