Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Ethoxycarbonylpropyl)triphenylphosphonium bromide, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
50479-11-3
C24H26BrO2P
457.348
MFCD00274610
JPZMNVPVVYVXAD-UHFFFAOYSA-M
3-ethoxycarbonyl propyl triphenylphosphonium bromide,4-ethoxy-4-oxobutyl triphenylphosphonium bromide,ethoxycarbonylpropyltriphenylphosphonium bromide,4-ethoxy-4-oxobutyl triphenylphosphanium bromide,phosphonium, 4-ethoxy-4-oxobutyl triphenyl-, bromide 1:1,acmc-1anq6,3-carboethoxypropyltriphenylphosphonium bromide,3-carboethoxypropyl triphenylphosphonium bromide,3-ethoxycarbonylpropyltriphenylphosphonium bromide,4-ethoxy-4-oxobutyl-triphenylphosphanium bromide
2773643
(4-ethoxy-4-oxobutyl)-triphenylphosphanium;bromide
CCOC(=O)CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
50479-11-3
C24H26BrO2P
457.348
MFCD00274610
JPZMNVPVVYVXAD-UHFFFAOYSA-M
3-ethoxycarbonyl propyl triphenylphosphonium bromide,4-ethoxy-4-oxobutyl triphenylphosphonium bromide,ethoxycarbonylpropyltriphenylphosphonium bromide,4-ethoxy-4-oxobutyl triphenylphosphanium bromide,phosphonium, 4-ethoxy-4-oxobutyl triphenyl-, bromide 1:1,acmc-1anq6,3-carboethoxypropyltriphenylphosphonium bromide,3-carboethoxypropyl triphenylphosphonium bromide,3-ethoxycarbonylpropyltriphenylphosphonium bromide,4-ethoxy-4-oxobutyl-triphenylphosphanium bromide
2773643
(4-ethoxy-4-oxobutyl)-triphenylphosphanium;bromide
CCOC(=O)CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
For C-C Bond Formation (Olefination)
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 167°C to 170°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 457.35
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material (3-Ethoxycarbonylpropyl)triphenylphosphonium bromide
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Contact with water liberates toxic gas.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning