Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Ethoxymethylene)-2,4-pentanedione, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
33884-41-2
C8H12O3
156.181
MFCD07368652
ITTXGKOHFZJUEX-UHFFFAOYSA-N
3-ethoxymethylene pentane-2,4-dione,3-ethoxymethylene-2,4-pentanedione,3-ethoxymethylidene pentane-2,4-dione,acmc-1cmxl,3-acetyl-4-ethoxy-3-buten-2-one,3-ethoxymethylene-pentan-2,4-dione,2,4-pentanedione,3-ethoxymethylene,2,4-pentanedione, 3-ethoxymethylene,methyl 1-methylcarbonyl-2-ethoxyvinyl ketone
240418
3-(ethoxymethylidene)pentane-2,4-dione
CCOC=C(C(=O)C)C(=O)C
33884-41-2
C8H12O3
156.181
MFCD07368652
ITTXGKOHFZJUEX-UHFFFAOYSA-N
3-ethoxymethylene pentane-2,4-dione,3-ethoxymethylene-2,4-pentanedione,3-ethoxymethylidene pentane-2,4-dione,acmc-1cmxl,3-acetyl-4-ethoxy-3-buten-2-one,3-ethoxymethylene-pentan-2,4-dione,2,4-pentanedione,3-ethoxymethylene,2,4-pentanedione, 3-ethoxymethylene,methyl 1-methylcarbonyl-2-ethoxyvinyl ketone
240418
3-(ethoxymethylidene)pentane-2,4-dione
CCOC=C(C(=O)C)C(=O)C
Packaging Glass bottle
Color Yellow
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CH2OCHC(C(O)CH3)2
Formula Weight 156.18
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-(Ethoxymethylene)-2, 4-pentanedione
Show More Show Less