Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Ethoxyphenol, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Boiling Point | 247°C |
Refractive Index | 1.541 |
Density | 1.07 |
Melting Point | 28°C to 31°C |
Flash Point | >93°C (199°F) |
Odor | Phenol-like |
Quantity | 25g |
Formula Weight | 138.17 |
Percent Purity | 98% |
Chemical Name or Material | 3-Ethoxyphenol |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 210-681-8
TSCA : Yes
Recommended Storage : Ambient temperatures