Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxyphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
90555-66-1
C8H11BO3
165.99
CHCWUTJYLUBETR-UHFFFAOYSA-N
3-ethoxyphenyl boronic acid,3-ethoxybenzeneboronic acid,3-ethoxyphenylboronicacid,3-ethoxyphenyl boranediol,m-ethoxyphenylboronic acid,boronic acid, 3-ethoxyphenyl,pubchem1835,acmc-209r6q,3-ethoxy-phenylboronic acid
2773920
(3-ethoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OCC)(O)O
90555-66-1
C8H11BO3
165.99
CHCWUTJYLUBETR-UHFFFAOYSA-N
3-ethoxyphenyl boronic acid,3-ethoxybenzeneboronic acid,3-ethoxyphenylboronicacid,3-ethoxyphenyl boranediol,m-ethoxyphenylboronic acid,boronic acid, 3-ethoxyphenyl,pubchem1835,acmc-209r6q,3-ethoxy-phenylboronic acid
2773920
(3-ethoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OCC)(O)O
Melting Point 151.5°C to 152.2°C
Quantity 5g
Infrared Spectrum Authentic
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 165.99
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Ethoxyphenylboronic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning