Alfa Aesar™ 3-Ethoxypropylamine, 99%

Manufacturer: Alfa Aesar™ B2369422

Quantity 100g

 View more versions of this product

Catalog No. AAB2369422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6291-85-6
    Molecular Formula C5H13NO
    Linear Formula CH3CH2O(CH2)3NH2
    Formula Weight 103.16
    MDL Number MFCD00008221
    UN Number UN2733
    Beilstein 1732287
    Chemical Name or Material 3-Ethoxypropylamine

    Specifications

    Formula Weight 103.16
    TSCA TSCA
    Linear Formula CH3CH2O(CH2)3NH2
    MDL Number MFCD00008221
    UN Number UN2733
    Beilstein 1732287
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    RTECS Number UI2700000
    Flash Point 32°C (91°F)
    Quantity 100g
    Molecular Formula C5H13NO
    CAS 6291-85-6
    Health Hazard 1 Danger
    Boiling Point 136° to 138°C
    Chemical Name or Material 3-Ethoxypropylamine
    Assay 99%

    Description

    Applications
    It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.

    Solubility
    Miscible in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit