Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Ethoxythiobenzamide, 97%

Manufacturer:  Alfa Aesar™ H5222703

Quantity 1g

 View more versions of this product

Catalog No. AAH5222703


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

747411-11-6
181.25
3-Ethoxythiobenzamide
C9H11NOS
MFCD05270860

Specifications

1g
C9H11NOS
MFCD05270860
97%
747411-11-6
181.25
3-Ethoxythiobenzamide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit