Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Ethyl-2-methylbenzoxazolium iodide, 95%

Click to view available options
Quantity:
5g
25g
100g
5260-37-7
C10H12INO
289.116
MFCD00044141
UZWYLWSUBQEUMC-UHFFFAOYSA-M
3-ethyl-2-methylbenzoxazolium iodide,3-ethyl-2-methylbenzo d oxazol-3-ium iodide,2-methyl-3-ethylbenzoxazolium iodide,benzoxazolium, 3-ethyl-2-methyl-, iodide,benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1,3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide,3-ethyl-2-methylbenzoxazoliumiodide,acmc-209l0h,ethyl-2-methyl-benzoxazolium iodide,3-ethyl-2-methylbenzoxazolium iodide, technical grade
21331
3-ethyl-2-methyl-1,3-benzoxazol-3-ium;iodide
CC[N+]1=C(OC2=CC=CC=C21)C.[I-]
5260-37-7
C10H12INO
289.116
MFCD00044141
UZWYLWSUBQEUMC-UHFFFAOYSA-M
3-ethyl-2-methylbenzoxazolium iodide,3-ethyl-2-methylbenzo d oxazol-3-ium iodide,2-methyl-3-ethylbenzoxazolium iodide,benzoxazolium, 3-ethyl-2-methyl-, iodide,benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1,3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide,3-ethyl-2-methylbenzoxazoliumiodide,acmc-209l0h,ethyl-2-methyl-benzoxazolium iodide,3-ethyl-2-methylbenzoxazolium iodide, technical grade
21331
3-ethyl-2-methyl-1,3-benzoxazol-3-ium;iodide
CC[N+]1=C(OC2=CC=CC=C21)C.[I-]
Sensitivity Hygroscopic; Light sensitive
Quantity 5g
Melting Point 196°C to 198°C (decomposition)
Formula Weight 289.12
Percent Purity 95%
Chemical Name or Material 3-Ethyl-2-methylbenzoxazolium iodide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-068-3

RTECSNumber : DM5285000

TSCA : Yes

Recommended Storage : Ambient temperatures