Alfa Aesar™ 3-Ethylphenol, 95%

Manufacturer: Alfa Aesar™ B2347018

Quantity 50g

 View more versions of this product

Catalog No. AAB2347018

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 620-17-7
    Molecular Formula C8H10O
    Formula Weight 122.17
    MDL Number MFCD00002311
    UN Number UN3145
    Beilstein 1857002
    Chemical Name or Material 3-Ethylphenol

    Specifications

    Formula Weight 122.17
    TSCA TSCA
    Flash Point 94°C (202°F)
    Boiling Point 218°C
    Quantity 50g
    Molecular Formula C8H10O
    MDL Number MFCD00002311
    CAS 620-17-7
    UN Number UN3145
    Beilstein 1857002
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.
    Melting Point −4°C
    Chemical Name or Material 3-Ethylphenol
    Assay 95%

    Description

    Applications
    It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

    Solubility
    Slightly soluble in water.

    Notes
    Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

    GHS H Statement
    H301-H311-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit