Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Ethylphenol, 95%

Click to view available options
Quantity:
10g
50g
250g
620-17-7
C8H10O
122.167
MFCD00002311
HMNKTRSOROOSPP-UHFFFAOYSA-N
m-ethylphenol,phenol, 3-ethyl,phenol, m-ethyl,meta-ethylphenol,1-ethyl-3-hydroxybenzene,1-hydroxy-3-ethylbenzene,3-ethyl-phenol,unii-0g9zk222jx,benzene, 1-ethyl-3-hydroxy,dsstox_cid_2480
12101
CHEBI:34332
3-ethylphenol
CCC1=CC(=CC=C1)O
620-17-7
C8H10O
122.167
MFCD00002311
HMNKTRSOROOSPP-UHFFFAOYSA-N
m-ethylphenol,phenol, 3-ethyl,phenol, m-ethyl,meta-ethylphenol,1-ethyl-3-hydroxybenzene,1-hydroxy-3-ethylbenzene,3-ethyl-phenol,unii-0g9zk222jx,benzene, 1-ethyl-3-hydroxy,dsstox_cid_2480
12101
CHEBI:34332
3-ethylphenol
CCC1=CC(=CC=C1)O
Applications
It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Melting Point -4°C
Boiling Point 218°C
Quantity 250g
Flash Point 94°C (202°F)
UN Number UN3145
Beilstein 1857002
Formula Weight 122.17
Percent Purity 95%
Chemical Name or Material 3-Ethylphenol

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.

TSCA : TSCA