Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethynylaniline 98.0+%, TCI America™

Click to view available options
Quantity:
10g
25g
54060-30-9
C8H7N
117.151
MFCD00014779
NNKQLUVBPJEUOR-UHFFFAOYSA-N
3-aminophenylacetylene,benzenamine, 3-ethynyl,m-aminophenylacetylene,3-ethynylbenzenamine,3-ethynylphenylamine,3-ethynylaniline,m-aminophenyl acetylene,3-aminophenyl acetylene,m-ethynylaniline,3-ethynyl-aniline
104682
3-ethynylaniline
C#CC1=CC(=CC=C1)N
54060-30-9
C8H7N
117.151
MFCD00014779
NNKQLUVBPJEUOR-UHFFFAOYSA-N
3-aminophenylacetylene,benzenamine, 3-ethynyl,m-aminophenylacetylene,3-ethynylbenzenamine,3-ethynylphenylamine,3-ethynylaniline,m-aminophenyl acetylene,3-aminophenyl acetylene,m-ethynylaniline,3-ethynyl-aniline
104682
3-ethynylaniline
C#CC1=CC(=CC=C1)N
Quantity 10g
Boiling Point 120°C
Color Brown
Formula Weight 117.15
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Ethynylaniline