Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-exo-(Benzyloxycarbonylamino)bicyclo]2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
109853-34-1
C16H17NO4
287.315
MFCD01863248
DGRPAZJZBQTEPY-DGAVXFQQSA-N
3-exo-benzyloxy carbonyl amino bicyclo 2.2.1 hept-5-ene-2-carboxylic acid,1r,4s-3beta-benzyloxycarbonylamino bicyclo 2.2.1 hepta-5-ene-2beta-carboxylic acid,bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-phenylmethoxy carbonyl amino-, exo,exo-9ci
7157280
(1R,2R,3S,4S)-3-(phenylmethoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
C1C2C=CC1C(C2C(=O)O)NC(=O)OCC3=CC=CC=C3
109853-34-1
C16H17NO4
287.315
MFCD01863248
DGRPAZJZBQTEPY-DGAVXFQQSA-N
3-exo-benzyloxy carbonyl amino bicyclo 2.2.1 hept-5-ene-2-carboxylic acid,1r,4s-3beta-benzyloxycarbonylamino bicyclo 2.2.1 hepta-5-ene-2beta-carboxylic acid,bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-phenylmethoxy carbonyl amino-, exo,exo-9ci
7157280
(1R,2R,3S,4S)-3-(phenylmethoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
C1C2C=CC1C(C2C(=O)O)NC(=O)OCC3=CC=CC=C3
Packaging Glass bottle
Melting Point 119°C to 123°C
Color Beige
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 287.31
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-exo-(Benzyloxycarbonylamino)bicyclo]2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%
Show More Show Less